Assay Details
Gene Symbol: DMWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839492.1 | XP_028695325.1 | 2050 | 86 | ||
GenBank mRNA | JV046347.1 | - | 4 - 5 | 1979 | 86 | |
JV667355.1 | - | 4 - 5 | 1975 | 86 | ||
JV708584.1 | - | 4 - 5 | 1797 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
