Assay Details
Gene Symbol: MIR654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032676.1 | 1 | 41 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | OTU deubiquitinase 7A | OTUD7A | Anti-sense | Chr. 7 - 6425374 - 6800098 on NCBI Build Mmul_10 | ||
Downstream | Kruppel like factor 13 | KLF13 | 280.8kb | Sense | Chr. 7 - 6888981 - 6939462 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
