Assay Details
Gene Symbol: SALL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126115.2 | XP_014981601.2 | 3854 | 79 | ||
GenBank mRNA | JU335839.1 | - | 2 - 3 | 3550 | 79 | |
JV720752.1 | - | 1 - 2 | 3044 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
