Assay Details
Gene Symbol: KCNMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002798660.3 | XP_002798706.2 | 896 | 105 | ||
GenBank mRNA | JU334041.1 | - | 389 | 105 | ||
JU475209.1 | - | 370 | 105 | |||
JV045550.1 | - | 381 | 105 | |||
JV613478.1 | - | 413 | 105 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
