Assay Details
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244590.2 | NP_001231519.1 | 221 | 66 | ||
GenBank mRNA | JU324924.1 | - | 198 | 66 | ||
JU472640.1 | - | 109 | 66 | |||
JV048768.1 | - | 187 | 66 | |||
JV601714.1 | - | 198 | 66 | |||
JV637187.1 | - | 192 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
