Assay Details
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194894.3 | NP_001181823.2 | 1 - 2 | 458 | 63 | |
XM_015143263.2 | XP_014998749.2 | 577 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
