Assay Details
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194894.3 | NP_001181823.2 | 3 - 4 | 757 | 70 | |
XM_015143263.2 | XP_014998749.2 | 876 | 70 | |||
GenBank mRNA | JU326004.1 | - | 2 - 3 | 240 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
