Assay Details
Gene Symbol: LITAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258181.1 | NP_001245110.1 | 412 | 113 | ||
GenBank mRNA | JU323150.1 | - | 452 | 113 | ||
JU472770.1 | - | 412 | 113 | |||
JV704028.1 | - | 455 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
