Assay Details
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260532.1 | NP_001247461.1 | 2 - 3 | 121 | 99 | |
XM_028843133.1 | XP_028698966.1 | 1312 | 99 | |||
XM_028843136.1 | XP_028698969.1 | 1231 | 99 | |||
XM_028843140.1 | XP_028698973.1 | 1073 | 99 | |||
GenBank mRNA | JU335182.1 | - | 2 - 3 | 156 | 99 | |
JV045515.1 | - | 2 - 3 | 121 | 99 | ||
JV654847.1 | - | 2 - 3 | 166 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
