Assay Details
Gene Symbol: MRPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257670.1 | NP_001244599.1 | 359 | 92 | ||
GenBank mRNA | JU335926.1 | - | 359 | 92 | ||
JU473679.1 | - | 449 | 92 | |||
JV044088.1 | - | 428 | 92 | |||
JV566998.1 | - | 449 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
