Assay Details
Gene Symbol: LAMTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257681.1 | NP_001244610.1 | 356 | 86 | ||
GenBank mRNA | JU320713.1 | - | 356 | 86 | ||
JU473738.1 | - | 395 | 86 | |||
JV043220.1 | - | 329 | 86 |
