Assay Details
Gene Symbol: LAMTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257681.1 | NP_001244610.1 | 3 - 4 | 452 | 115 | |
GenBank mRNA | JU320713.1 | - | 3 - 4 | 452 | 115 | |
JU473738.1 | - | 3 - 4 | 491 | 115 | ||
JV043220.1 | - | 3 - 4 | 425 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
