Assay Details
Gene Symbol: DDX46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261180.1 | NP_001248109.1 | 2950 | 124 | ||
XM_015140860.2 | XP_014996346.1 | 3041 | 124 | |||
GenBank mRNA | JU337370.1 | - | 2981 | 124 | ||
JU478067.1 | - | 2931 | 124 | |||
JV048852.1 | - | 2950 | 124 | |||
JV599884.1 | - | 3005 | 124 |
Gene Symbol: LOC107000127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147052.2 | XP_015002538.2 | 2578 | 124 |

More Information
Set Membership: |
![]() ![]() |
