Assay Details
Gene Symbol: DDX46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261180.1 | NP_001248109.1 | 3161 | 82 | ||
XM_015140860.2 | XP_014996346.1 | 3252 | 82 | |||
GenBank mRNA | JU337370.1 | - | 3192 | 82 | ||
JU478067.1 | - | 3142 | 82 | |||
JV048852.1 | - | 3161 | 82 | |||
JV599884.1 | - | 3216 | 82 |
Gene Symbol: LOC107000127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147052.2 | XP_015002538.2 | 2789 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
