Assay Details
Gene Symbol: MALSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261201.2 | NP_001248130.1 | 1 - 2 | 271 | 84 | |
GenBank mRNA | JU332817.1 | - | 1 - 2 | 266 | 84 | |
JU474229.1 | - | 1 - 2 | 271 | 84 | ||
JV046123.1 | - | 1 - 2 | 260 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
