Assay Details
Gene Symbol: NUDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261225.1 | NP_001248154.1 | 339 | 106 | ||
XM_028845013.1 | XP_028700846.1 | 563 | 106 | |||
GenBank mRNA | JU322807.1 | - | 339 | 106 | ||
JU478156.1 | - | 351 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
