Assay Details
Gene Symbol: ISYNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261319.1 | NP_001248248.1 | 7 - 8 | 1036 | 62 | |
XM_015123293.2 | XP_014978779.2 | 1168 | 62 | |||
XM_028838525.1 | XP_028694358.1 | 1730 | 62 | |||
XM_028838526.1 | XP_028694359.1 | 1631 | 62 | |||
GenBank mRNA | JU334903.1 | - | 7 - 8 | 1052 | 62 | |
JU475908.1 | - | 7 - 8 | 1036 | 62 | ||
JV047308.1 | - | 7 - 8 | 1073 | 62 | ||
JV686389.1 | - | 7 - 8 | 1022 | 62 |
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV047307.1 | - | 7 - 8 | 1073 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
