Assay Details
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261474.1 | NP_001248403.1 | 474 | 57 | ||
GenBank mRNA | JU333707.1 | - | 474 | 57 | ||
JV576687.1 | - | 514 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
