Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261483.1 | NP_001248412.1 | 320 | 110 | ||
GenBank mRNA | JU332962.1 | - | 320 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
