Assay Details
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265666.1 | NP_001252595.1 | 1 - 2 | 838 | 67 | |
XM_015121917.2 | XP_014977403.1 | 1772 | 67 | |||
XM_015121918.2 | XP_014977404.1 | 1580 | 67 | |||
XM_015121919.2 | XP_014977405.1 | 1684 | 67 | |||
XM_015121920.2 | XP_014977406.1 | 1605 | 67 | |||
XM_015121921.2 | XP_014977407.1 | 1648 | 67 | |||
XM_028837715.1 | XP_028693548.1 | 1865 | 67 | |||
XM_028837716.1 | XP_028693549.1 | 1829 | 67 | |||
XM_028837717.1 | XP_028693550.1 | 1621 | 67 | |||
XM_028837718.1 | XP_028693551.1 | 1862 | 67 | |||
GenBank mRNA | JV566742.1 | - | 2 - 3 | 1545 | 67 | |
JV635691.1 | - | 1 - 2 | 838 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
