Assay Details
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266453.1 | NP_001253382.1 | 3 - 4 | 395 | 96 | |
XM_028849143.1 | XP_028704976.1 | 416 | 96 | |||
GenBank mRNA | JU334426.1 | - | 3 - 4 | 394 | 96 | |
JV045084.1 | - | 3 - 4 | 395 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
