Assay Details
Gene Symbol: UNC119B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266482.1 | NP_001253411.1 | 668 | 93 | ||
GenBank mRNA | JV043636.1 | - | 651 | 93 | ||
JV635687.1 | - | 668 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
