Assay Details
Gene Symbol: BTN2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266531.1 | NP_001253460.1 | 6 - 7 | 1040 | 101 | |
XM_015135627.2 | XP_014991113.2 | 1069 | 101 | |||
XM_015135628.2 | XP_014991114.2 | 1101 | 101 | |||
XM_015135629.2 | XP_014991115.2 | 1174 | 101 | |||
GenBank mRNA | JU473960.1 | - | 6 - 7 | 1040 | 101 | |
JU473961.1 | - | 6 - 7 | 1040 | 101 | ||
JU473963.1 | - | 6 - 7 | 1040 | 101 | ||
JU473964.1 | - | 6 - 7 | 1040 | 101 | ||
JU473966.1 | - | 6 - 7 | 1040 | 101 | ||
JU473967.1 | - | 6 - 7 | 1040 | 101 | ||
JU473968.1 | - | 6 - 7 | 1040 | 101 |
Gene Symbol: LOC705417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146564.2 | XP_015002050.2 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
