Assay Details
Gene Symbol: LPAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266718.1 | NP_001253647.1 | 870 | 85 | ||
XM_028838546.1 | XP_028694379.1 | 797 | 85 | |||
GenBank mRNA | JU477801.1 | - | 860 | 85 | ||
JV043877.1 | - | 844 | 85 | |||
JV636448.1 | - | 870 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
