Assay Details
Gene Symbol: PRXL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002805660.3 | XP_002805706.2 | 345 | 75 | ||
XM_015147134.2 | XP_015002620.1 | 362 | 75 | |||
XM_015147135.2 | XP_015002621.1 | 397 | 75 | |||
XM_015147136.2 | XP_015002622.1 | 259 | 75 | |||
XM_028826411.1 | XP_028682244.1 | 253 | 75 | |||
XM_028826412.1 | XP_028682245.1 | 190 | 75 | |||
GenBank mRNA | JU473710.1 | - | 2 - 3 | 305 | 75 | |
JU473711.1 | - | 2 - 3 | 305 | 75 | ||
JV045354.1 | - | 2 - 3 | 304 | 75 | ||
JV635801.1 | - | 2 - 3 | 289 | 75 | ||
JV668307.1 | - | 2 - 3 | 289 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
