Assay Details
Gene Symbol: C1QTNF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139805.2 | XP_014995291.2 | 658 | 110 | ||
GenBank mRNA | JV651026.1 | - | 2 - 3 | 198 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
