Assay Details
Gene Symbol: GAREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111866.2 | XP_014967352.2 | 423 | 66 | ||
XM_015111867.2 | XP_014967353.2 | 302 | 66 | |||
GenBank mRNA | JV674109.1 | - | 353 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015111867.2 transcripts.
Set Membership: |
![]() ![]() |
