Assay Details
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002805559.3 | XP_002805605.1 | 998 | 91 | ||
XM_002805561.3 | XP_002805607.1 | 848 | 91 | |||
XM_015146520.2 | XP_015002006.1 | 834 | 91 | |||
XM_015146524.2 | XP_015002010.1 | 1170 | 91 | |||
XM_028826006.1 | XP_028681839.1 | 1664 | 91 | |||
XM_028826007.1 | XP_028681840.1 | 1010 | 91 | |||
GenBank mRNA | JU320046.1 | - | 825 | 91 | ||
JU320047.1 | - | 825 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
