Assay Details
Gene Symbol: PET100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097045.3 | XP_001097045.1 | 116 | 81 | ||
GenBank mRNA | JU332371.1 | - | 1 - 2 | 70 | 81 | |
JV045114.1 | - | 1 - 2 | 79 | 81 | ||
JV491178.1 | - | 1 - 2 | 75 | 81 | ||
JV601819.1 | - | 1 - 2 | 49 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001097045.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
