Assay Details
Gene Symbol: SEMG1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038076.2 | 3 - 4 | 1502 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | synapsin III | SYN3 | -76.3kb | Anti-sense | Chr. 10 - 18451918 - 19008993 on NCBI Build Mmul_10 | |
Downstream | RNA 2',3'-cyclic phosphate and 5'-OH ligase | RTCB |
C10H22orf28, C22orf28, EGK_02972 |
8.9kb | Anti-sense | Chr. 10 - 19096161 - 19122161 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
