Assay Details
Gene Symbol: ASPHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149964.2 | XP_015005450.1 | 1441 | 69 | ||
GenBank mRNA | JU335587.1 | - | 1024 | 69 | ||
JU473560.1 | - | 1162 | 69 | |||
JV045183.1 | - | 1314 | 69 | |||
JV565730.1 | - | 1388 | 69 | |||
JV704051.1 | - | 1309 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
