Assay Details
Gene Symbol: ADA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149658.2 | XP_015005144.2 | 1145 | 108 | ||
XM_015149659.2 | XP_015005145.2 | 1048 | 108 | |||
GenBank mRNA | JU475991.1 | - | 4 - 5 | 938 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
