Assay Details
Gene Symbol: ADA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149658.2 | XP_015005144.2 | 1230 | 106 | ||
XM_015149659.2 | XP_015005145.2 | 1133 | 106 | |||
GenBank mRNA | JU475991.1 | - | 5 - 6 | 1023 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
