Assay Details
Gene Symbol: ADA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149658.2 | XP_015005144.2 | 1502 | 64 | ||
XM_015149659.2 | XP_015005145.2 | 1405 | 64 | |||
GenBank mRNA | JU475991.1 | - | 7 - 8 | 1295 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
