Assay Details
Gene Symbol: BICRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840004.1 | XP_028695837.1 | 352 | 70 | ||
XM_028840005.1 | XP_028695838.1 | 490 | 70 | |||
XM_028840006.1 | XP_028695839.1 | 352 | 70 | |||
GenBank mRNA | JU485284.1 | - | 194 | 70 | ||
JV614748.1 | - | 198 | 70 |

More Information
Set Membership: |
![]() ![]() |
