Assay Details
Gene Symbol: XRCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108141.4 | XP_001108141.2 | 242 | 114 | ||
GenBank mRNA | JU477096.1 | - | 2 - 3 | 201 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
