Assay Details
Gene Symbol: TTLL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150607.2 | XP_015006093.2 | 1428 | 68 | ||
XM_028827569.1 | XP_028683402.1 | 1132 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
