Assay Details
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112523.2 | XP_014968009.1 | 692 | 61 | ||
XM_015112529.2 | XP_014968015.1 | 525 | 61 | |||
GenBank mRNA | JU333365.1 | - | 582 | 61 | ||
JU474659.1 | - | 501 | 61 | |||
JV046434.1 | - | 699 | 61 | |||
JV645040.1 | - | 680 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
