Assay Details
Gene Symbol: CCND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278446.1 | NP_001265375.1 | 778 | 59 | ||
GenBank mRNA | JU335774.1 | - | 722 | 59 | ||
JU473363.1 | - | 719 | 59 | |||
JV515827.1 | - | 778 | 59 | |||
JV589729.1 | - | 765 | 59 | |||
JV720426.1 | - | 722 | 59 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
