Assay Details
Gene Symbol: GNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120126.2 | XP_014975612.1 | 839 | 103 | ||
XM_015120128.2 | XP_014975614.1 | 450 | 103 | |||
GenBank mRNA | JU320394.1 | - | 601 | 103 | ||
JU320395.1 | - | 601 | 103 | |||
JU320396.1 | - | 601 | 103 | |||
JU474043.1 | - | 620 | 103 | |||
JU474044.1 | - | 620 | 103 | |||
JU474045.1 | - | 620 | 103 | |||
JV043386.1 | - | 560 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
