Assay Details
Gene Symbol: SEPTIN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120460.2 | XP_014975946.2 | 1308 | 91 | ||
XM_015120462.2 | XP_014975948.2 | 1032 | 91 | |||
XM_015120463.2 | XP_014975949.2 | 1470 | 91 | |||
XM_015120464.2 | XP_014975950.2 | 1692 | 91 | |||
XM_015120465.2 | XP_014975951.1 | 447 | 91 | |||
XM_015120466.2 | XP_014975952.1 | 742 | 91 | |||
XM_015120467.2 | XP_014975953.1 | 551 | 91 | |||
XM_015120468.2 | XP_014975954.1 | 344 | 91 | |||
XM_028836849.1 | XP_028692682.1 | 777 | 91 | |||
XM_028836850.1 | XP_028692683.1 | 282 | 91 | |||
GenBank mRNA | JU322456.1 | - | 3 - 4 | 1035 | 91 | |
JV045987.1 | - | 3 - 4 | 1092 | 91 | ||
JV527780.1 | - | 3 - 4 | 936 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
