Assay Details
Gene Symbol: TGFBR3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839638.1 | XP_028695471.1 | 294 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
