Assay Details
Gene Symbol: CBARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028838766.1 | XP_028694599.1 | 92 | |||
XM_028838767.1 | XP_028694600.1 | 92 | ||||
XM_028838768.1 | XP_028694601.1 | 92 | ||||
GenBank mRNA | JU329207.1 | - | 283 | 92 | ||
JV577254.1 | - | 378 | 92 | |||
JV679138.1 | - | 380 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
