Assay Details
Gene Symbol: CBARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028838766.1 | XP_028694599.1 | 1653 | 102 | ||
XM_028838767.1 | XP_028694600.1 | 1259 | 102 | |||
XM_028838768.1 | XP_028694601.1 | 1290 | 102 | |||
GenBank mRNA | JU329207.1 | - | 950 | 102 | ||
JV577254.1 | - | 1045 | 102 | |||
JV679138.1 | - | 1047 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
