Assay Details
Gene Symbol: ATP5PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260679.2 | NP_001247608.1 | 660 | 69 | ||
XM_028840436.1 | XP_028696269.1 | 575 | 69 | |||
GenBank mRNA | JU334908.1 | - | 630 | 69 | ||
JU475913.1 | - | 525 | 69 | |||
JV044839.1 | - | 661 | 69 | |||
JV470977.1 | - | 625 | 69 | |||
JV667200.1 | - | 591 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
