Assay Details
Gene Symbol: NUDT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015121054.2 | XP_014976540.2 | 533 | 105 | ||
GenBank mRNA | JU476252.1 | - | 379 | 105 | ||
JV047540.1 | - | 469 | 105 | |||
JV047541.1 | - | 469 | 105 | |||
JV636253.1 | - | 442 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
