Assay Details
Gene Symbol: MIR28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031998.1 | 1 | 47 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cell adhesion molecule 2 | CADM2 | EGK_11194 | -8.9kb | Anti-sense | Chr. 2 - 185956279 - 187037122 on NCBI Build Mmul_10 |
Downstream | 1,4-alpha-glucan branching enzyme 1 | GBE1 | EGK_11193 | 3.2mb | Sense | Chr. 2 - 190229211 - 190491925 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
