Assay Details
Gene Symbol: MIR221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032052.1 | 1 | 54 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | p21 (RAC1) activated kinase 3 | PAK3 | Sense | Chr. X - 107644668 - 107937809 on NCBI Build Mmul_10 | ||
Downstream | calpain 6 | CAPN6 |
EGK_20831, calpain-6 |
262.6kb | Anti-sense | Chr. X - 107959423 - 107984698 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
