Assay Details
Gene Symbol: MIR373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032495.1 | 1 | 37 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | fem-1 homolog A | FEM1A | -30.5kb | Anti-sense | Chr. 19 - 4725656 - 4728461 on NCBI Build Mmul_10 | |
Downstream | perilipin 3 | PLIN3 | 13.5kb | Sense | Chr. 19 - 4772430 - 4804130 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
