Assay Details
Gene Symbol: MIR550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032604.1 | 1 | 43 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | diacylglycerol kinase beta | DGKB | Sense | Chr. 3 - 100092594 - 100860322 on NCBI Build Mmul_10 | ||
Downstream |
myosin, light chain 6, alkali, smooth muscle and non-muscle-like |
LOC710137 | 279.5kb | Sense | Chr. 3 - 100757264 - 100757983 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
